0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Austria
ContactUsLinkComponent
Proteins and Peptides
Recombinant Hepatitis A Virus P3C
Recombinant Hepatitis A Virus P3C
Recombinant Hepatitis A Virus P3C
Catalog # APOSBITP1817-1MG
Supplier:
Apollo Scientific
CAS Number:
Expand image
Recombinant Hepatitis A Virus P3C
Catalog # APOSBITP1817-1MG
Supplier:
Apollo Scientific
CAS Number:
Specifications
Pk:
1 mg
Protein/peptide type:
Recombinant
Protein synonyms:
HAV P3C
Protein/peptide name:
Hepatitis A Virus P3C
Specifications
About this item
Product Details
Product Details