Order Entry
Austria
ContactUsLinkComponent
Recombinant Hepatitis A Virus VP1-P2A (722-830)
Recombinant Hepatitis A Virus VP1-P2A (722-830)
Catalog # APOSBITP1812-1MG
Supplier:  Apollo Scientific
CAS Number:  
Recombinant Hepatitis A Virus VP1-P2A (722-830)
Catalog # APOSBITP1812-1MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Protein/peptide type:
    Recombinant
  • Protein synonyms:
    HAV VP1-P2A
  • Protein/peptide name:
    Hepatitis A Virus VP1-P2A (722-830)

Specifications

About this item