Order Entry
Austria
ContactUsLinkComponent
Recombinant Hepatitis C Virus NS4
Recombinant Hepatitis C Virus NS4
Catalog # APOSBITP1788-1MG
Supplier:  Apollo Scientific
CAS Number:  
Recombinant Hepatitis C Virus NS4
Catalog # APOSBITP1788-1MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Protein/peptide type:
    Recombinant
  • Protein synonyms:
    HCV NS4
  • Protein/peptide name:
    Hepatitis C Virus NS4

Specifications

About this item