0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Austria
ContactUsLinkComponent
Proteins and Peptides
Recombinant Mosaic Hepatitis C Virus NS4 Genotype-2
Recombinant Mosaic Hepatitis C Virus NS4 Genotype-2
APOSBITP1785-1MG
:
Apollo Scientific
:
Recombinant Mosaic Hepatitis C Virus NS4 Genotype-2
APOSBITP1785-1MG
:
Apollo Scientific
:
Pk:
1 mg
Protein/peptide type:
Recombinant
Protein synonyms:
HCV NS4 Genotype-2
Protein/peptide name:
Mosaic Hepatitis C Virus NS4 Genotype-2