Order Entry
Austria
ContactUsLinkComponent
 
Recombinant Hepatitis C Virus NS3 Genotype-6 (1356-1459)
Catalog # APOSBITP1779-1MG
Supplier:  Apollo Scientific
CAS Number:  
Recombinant Hepatitis C Virus NS3 Genotype-6 (1356-1459)
Catalog # APOSBITP1779-1MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Protein/peptide type:
    Recombinant
  • Protein synonyms:
    HCV NS3 Genotype-6
  • Protein/peptide name:
    Hepatitis C Virus NS3 Genotype-6 (1356-1459)

Specifications

About this item