Order Entry
Austria
ContactUsLinkComponent
Recombinant Hepatitis C Virus NS3 Genotype-1a (1192-1459)
Recombinant Hepatitis C Virus NS3 Genotype-1a (1192-1459)
Catalog # APOSBITP1764-1MG
Supplier:  Apollo Scientific
CAS Number:  
Recombinant Hepatitis C Virus NS3 Genotype-1a (1192-1459)
Catalog # APOSBITP1764-1MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Protein/peptide type:
    Recombinant
  • Protein synonyms:
    HCV NS3 Genotype-1a
  • Protein/peptide name:
    Hepatitis C Virus NS3 Genotype-1a (1192-1459)

Specifications

About this item