Recombinant Hepatitis C Virus Nucleocapsid (core) 22kDa, Fluorescein
- Conjugation:Fluorescein
- Protein/peptide type:Recombinant
- Protein synonyms:HCV 22kDa
- Protein/peptide name:Hepatitis C Virus Nucleocapsid (core) 22kDa
Frequently Bought Together