Order Entry
Austria
ContactUsLinkComponent
Recombinant Borrelia p41
Recombinant Borrelia p41
Catalog # APOSBITP1735-1MG
Supplier:  Apollo Scientific
CAS Number:  
Recombinant Borrelia p41
Catalog # APOSBITP1735-1MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Protein/peptide type:
    Recombinant
  • Protein/peptide name:
    Borrelia p41

Specifications

About this item