Order Entry
Austria
ContactUsLinkComponent
Human recombinant alpha-synuclein
Human recombinant alpha-synuclein
Catalog # APOSBITP1463-1MG
Supplier:  Apollo Scientific
CAS Number:  
Human recombinant alpha-synuclein
Catalog # APOSBITP1463-1MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Protein synonyms:
    a-Synuclein
  • Protein/peptide name:
    Alpha-Synuclein

Specifications

About this item