Order Entry
Austria
ContactUsLinkComponent
Rabbit Recombinant Prl
Rabbit Recombinant Prl
Catalog # APOSBITP1249-1MG
Supplier:  Apollo Scientific
CAS Number:  
Rabbit Recombinant Prl
Catalog # APOSBITP1249-1MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Protein/peptide type:
    Recombinant
  • Species:
    Rabbit
  • Protein synonyms:
    Prolactin
  • Protein/peptide name:
    Prl

Specifications

About this item