Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Proteins and Peptides
Bovine Recombinant PL
Bovine Recombinant PL
APOSBITP1240-1MG
:
Apollo Scientific
:
Bovine Recombinant PL
APOSBITP1240-1MG
:
Apollo Scientific
:
Pk:
1 mg
Protein/peptide type:
Recombinant
Species:
Bovine
Protein synonyms:
Placental Lactogen
Protein/peptide name:
PL