Human Recombinant LFA-3
Catalog # APOSBITP1214-1MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pk:1 mg
- Protein/peptide type:Recombinant
- Species:Human
- Protein synonyms:Leukocyte Function Associated Antigen-3 Fusion Protein
- Protein/peptide name:LFA-3