Order Entry
Austria
ContactUsLinkComponent
Human Recombinant Leptin qA
Human Recombinant Leptin qA
Catalog # APOSBITP1206-1MG
Supplier:  Apollo Scientific
CAS Number:  
Human Recombinant Leptin qA
Catalog # APOSBITP1206-1MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Protein synonyms:
    Leptin Quadruple Antagonist
  • Protein/peptide name:
    Leptin qA

Specifications

About this item