Order Entry
Austria
ContactUsLinkComponent
Carp recombinant GH
Carp recombinant GH
Catalog # APOSBITP1077-1MG
Supplier:  Apollo Scientific
CAS Number:  
Carp recombinant GH
Catalog # APOSBITP1077-1MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Protein/peptide type:
    Recombinant
  • Species:
    Carp
  • Protein synonyms:
    Growth Hormone
  • Protein/peptide name:
    GH

Specifications

About this item