0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Austria
ContactUsLinkComponent
Proteins and Peptides
Sheep recombinant GH
Sheep recombinant GH
APOSBITP1075-1MG
:
Apollo Scientific
:
Sheep recombinant GH
APOSBITP1075-1MG
:
Apollo Scientific
:
Pk:
1 mg
Protein/peptide type:
Recombinant
Species:
Sheep
Protein synonyms:
Growth Hormone
Protein/peptide name:
GH