Order Entry
Austria
ContactUsLinkComponent
Human recombinant GLP-1
Human recombinant GLP-1
Catalog # APOSBITH3019-1MG
Supplier:  Apollo Scientific
CAS Number:  
Human recombinant GLP-1
Catalog # APOSBITH3019-1MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Protein synonyms:
    Glucagon Like Peptide-1 (7-36)
  • Protein/peptide name:
    GLP-1

Specifications

About this item