Order Entry
Austria
ContactUsLinkComponent
Mouse recombinant SDF-1b
Mouse recombinant SDF-1b
Catalog # APOSBITC2047-1MG
Supplier:  Apollo Scientific
CAS Number:  
Mouse recombinant SDF-1b
Catalog # APOSBITC2047-1MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Protein/peptide type:
    Recombinant
  • Species:
    Mouse
  • Protein synonyms:
    Stromal Cell-Derived Factor-1 beta, CXCL12
  • Protein/peptide name:
    SDF-1b

Specifications

About this item