Human recombinant MIP-4
Catalog # APOSBITC2040-1MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pk:1 mg
- Protein/peptide type:Recombinant
- Species:Human
- Protein synonyms:Macrophage Inflammatory protein-4 , CCL18
- Protein/peptide name:MIP-4