Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Proteins and Peptides
Human recombinant I-TAC
Human recombinant I-TAC
Human recombinant I-TAC
Catalog # APOSBITC2021-1MG
Supplier:
Apollo Scientific
CAS Number:
Expand image
Human recombinant I-TAC
Catalog # APOSBITC2021-1MG
Supplier:
Apollo Scientific
CAS Number:
Specifications
Pk:
1 mg
Protein/peptide type:
Recombinant
Species:
Human
Protein synonyms:
CXCL11
Protein/peptide name:
I-TAC
Specifications
About this item
Product Details
Product Details