Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Proteins and Peptides
Human recombinant GRO-a
Human recombinant GRO-a
Human recombinant GRO-a
Catalog # APOSBITC2010-1MG
Supplier:
Apollo Scientific
CAS Number:
Expand image
Human recombinant GRO-a
Catalog # APOSBITC2010-1MG
Supplier:
Apollo Scientific
CAS Number:
Specifications
Pk:
1 mg
Protein/peptide type:
Recombinant
Species:
Human
Seq:
23
Protein synonyms:
GRO-alpha, CXCL1
Protein/peptide name:
GRO-a
Specifications
About this item
Product Details
Product Details