Human recombinant IL-8-72
Catalog # APOSBITC2006-1MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pk:1 mg
- Protein/peptide type:Recombinant
- Species:Human
- Seq:23
- Protein synonyms:Interleukin-8, 1-72, CXCL8
- Protein/peptide name:IL-8-72