Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Proteins and Peptides
Fumonisin B3 (from
Fusarium moniliforme
)
Fumonisin B3 (from Fusarium moniliforme)
Fumonisin B3 (from Fusarium moniliforme)
Catalog # APOSBIF1008-1MG
Supplier:
Apollo Scientific
CAS Number:
Expand image
Fumonisin B3 (from Fusarium moniliforme)
Catalog # APOSBIF1008-1MG
Supplier:
Apollo Scientific
CAS Number:
Specifications
Pk:
1 mg
Source:
Fusarium moniliforme
Seq:
23
Protein/peptide name:
Fumonisin B3
Specifications
About this item
Product Details
Product Details