Order Entry
Austria
ContactUsLinkComponent
Human recombinant EGF (from Pichia pastoris)
Human recombinant EGF (from Pichia pastoris)
Supplier:  Apollo Scientific
Certificates
Human recombinant EGF (from Pichia pastoris)
Supplier:  Apollo Scientific

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    P. pastoris
  • Species:
    Human
  • Protein synonyms:
    Epidermal Growth Factor, Human, Recombinant
  • Protein/peptide name:
    EGF

Specifications

Frequently Bought Together