Gamma irradiated cleanroom markers, Sharpie®
These pens and markers eliminate the risks associated with surface sanitisation procedures.
- Lot controlled
- Ready to use
- Packed in easy to open Tyvek® peel pouches
- Minimises contamination risks
- Simplifies conformance to SOP's
- Eliminates the need for wiping down contaminated writing instruments
CRP offers gamma irradiated pens and markers for the convenience of pharmaceutical cleanroom operations. These are a 'Ready to Use Option' when sterilisation on-site is not possible; or surface sterilisation is insufficient. The clean Tyvek® peel pouch packaging allows for effective surface decontamination and easy introduction into the cleanroom through the material handling airlock.
: Certificates of Irradiation and Conformance with each shipment.
Frequently Bought Together