NANOCOLOR® Photometric Tube Test Kits for TOC
Supplier: MACHEREY-NAGEL
New Product
Certificates
About this item
A pre-dosed NANOCOLOR® tube test for photometric determination of TOC (total organic carbon) in water.
Measurement directly in the test tube, saving time and reducing exposure to chemical reagents
- Barcode cuvette identification
- Precisely pre-dosed reagents in 16 mm (ext.Ø) tubes
The test kits are evaluated in 16 mm (ext.Ø) tubes. They are preprogrammend on NANOCOLOR® photometers. Programming data for other spectrophotometers are available.
Sea water analysis: No
Hazardous material: Yes
Remark: Measuring range on NANOCOLOR VIS II
Sludge reagent set: No
Digestion set: No
NanOx Metal: No
NanOx N: No
Digestion temperature: 100 °C
Delivery: 3 rugged boxes with 60 test tubes. Sufficient for 20 tests.
Specifications
- Model:MANA985075
- Platform:NANOCOLOR® tube tests
- Storage temperature:2...8 °C
- Shelf life (months):12
- Weight:1054 g (Including packaging)
Specifications
Related Information
Certificates
Search by Lot NumberIFU NANOCOLOR® Photometric Tube Test Kits for TOC 30.pdf IFU NANOCOLOR® Photometric Tube Test Kits for TOC 300.pdf IFU NANOCOLOR® Photometric Tube Test Kits for TOC.pdf
Manual NANOCOLOR® Photometric Tube Test Kits for TOC 30.pdf Manual NANOCOLOR® Photometric Tube Test Kits for TOC 300.pdf
Manual
Photometric Tube Tests, NANOCOLOR® TOC 30.pdf
Additional Product Details
Frequently Bought Together