ContactUsLinkComponent
Coronavirus and SARS-CoV-2 ELISA kits, GSD NovaLisa®
Coronavirus and SARS-CoV-2 ELISA kits, GSD NovaLisa®
Catalog # GSDCCVGQ0970
CAS Number:  
undefined
Coronavirus and SARS-CoV-2 ELISA kits, GSD NovaLisa®
Catalog # GSDCCVGQ0970
CAS Number:  
Specifications
  • Pk:
    96 Tests
  • Description:
    GSD NovaLisa® SARS-CoV-2 (COVID-19) quantitative IgG
  • Seq:
    3
  • For:
    ThunderBolt® ELISA Analyser
  • Tests per kit:
    1×96 reactions
Specifications
About this item
Immunoassay for the quantitative and qualitative detection of specific IgG-class antibodies to the spike protein of SARS-CoV-2.
  • Quantitative determination of antibodies to the SARS-CoV-2 spike (S) protein
  • Trimeric SARS-CoV-2 Spike antigen
  • Intended as an aid to evaluate patients immune status and determine vaccine efficacy
  • Calibrators correlate to First WHO International Standards for SARS-CoV-2
  • Diagnostic sensitivity and specificity of 99.94% and 99.49% respectively
  • Validated for serum, plasma, and finger prick (serum from capillary blood) samples
It is intended to aid in the diagnosis of patients suspected of having COVID-19 disease or asymptomatic presentation of SARS-CoV-2, to monitor antibody levels during and after a COVID-19 infection, and to determine antibody levels before and after a COVID-19 vaccination.