Coronavirus and SARS-CoV-2 RT real-time PCR kits, GSD NovaPrime®
Supplier: GOLD STANDARD DIAGNOSTICS
Certificates
About this item
This RT-PCR assay is a highly sensitive Multiplex Real-Time PCR with CE-IVD marking for the direct qualitative pathogen detection of SARS-CoV-2 genomic RNA in respiratory samples.
- Approximately 1 hour turnaround time
- Specimen types: nasal wash/swab, nasopharyngeal wash/swab, oropharyngeal swab, BAL, pharynx gargle solution, and saliva
- Excellent performance with a 100% NPA, PPA
- Extraction/inhibition control included
The assay simultaneously detects two target sequences in the N gene.
Certifications: CE-IVD marked.
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together