Order Entry
Austria
ContactUsLinkComponent
 
NANOCOLOR® Photometric Tube Tests, Methanol
Catalog # MANA985859
Supplier:  MACHEREY-NAGEL
CAS Number:  
undefined
NANOCOLOR® Photometric Tube Tests, Methanol
Catalog # MANA985859
Supplier:  MACHEREY-NAGEL
CAS Number:  

Specifications

  • Chemical method:
    Alcoholoxidase/Peroxidase
  • Pk:
    1 KIT
  • Description:
    NANOCOLOR® Methanol 15, test no.8-59
  • Dimensions:
    160×110×250 mm (including packaging)
  • Seq:
    0
  • Platform:
    NANOCOLOR® tube tests
  • Storage temperature:
    <0 °C
  • Shelf life (months):
    24
  • Weight:
    573 g (including packaging)
  • Type:
    Hazardous material: No
  • For unit:
    UV/VIS II; VIS II; 500 D; PF-12Plus; Advance
  • Tests per kit:
    23
  • Range:
    0,2 - 15,0 mg/L (MeOH)

Specifications

About this item

Photometric test kit for the determination of methanol in water samples.

  • Measurement directly in the test tube, saving time and reducing exposure to chemical reagents
  • Bar code cuvette identification for easy test call up in NANOCOLOR® spectrophotometers
  • Precisely pre-dosed reagents with constant high quality from batch to batch
  • Colored pictogram instructions and easy handling of the tubes
  • 16 mm (OD) tubes for easy pipetting

The test kit is evaluated in 16 mm (AD) round cells. It is pre-programmed on the NANOCOLOR® photometers. Programming data for other spectrophotometers are available.

Sludge reagent set: No
Digestion set: No
NanOx Metal: No
NanOx N: No
Digestion temperature:

Delivery: Remark: Measuring range on NANOCOLOR VIS II