NANOCOLOR® Photometric Tube Test Kit for Ethanol
MANA985838
:
- Chemical method:Alcoholoxidase/Peroxidase
- Pk:1 KIT
- Description:NANOCOLOR® Ethanol 1000, test no. 8-38
- Dimensions:160×110×250 mm (including packaging)
- Model:MANA985838
- Platform:NANOCOLOR® tube tests
- Storage temperature:<0 °C
- Shelf life (months):24
- Weight:600 g including packaging
- For unit:UV/VIS II, VIS II, 500 D, PF-12Plus, Advance
- Tests per kit:23
- Range:0,1 - 1,00 g/L (EtOH); 0,013 - 0,130 Vol,% (EtOH)
Photometric test kit for the determination of ethanol in water samples.
- Measurement directly in the test tube, saving time and reducing exposure to chemical reagents Bar code cuvette identification for easy test call up in NANOCOLOR® spectrophotometers Precisely pre-dosed reagents with constant high quality from batch to batch Coloured pictogram instructions and easy handling of the tubes 16 mm (OD) tubes for easy pipetting
The test kit is evaluated in 16 mm (OD) tubes. It is preprogrammend on NANOCOLOR® photometers. Programming data for other spectrophotometers are available.
Sea water analysis: No Hazardous material: No Remark: Measuring range on NANOCOLOR VIS II
: Supplied in rugged box with 23 test tubes.