Order Entry
Austria
ContactUsLinkComponent
NANOCOLOR® Photometric Tube Test Kit for Chromate
NANOCOLOR® Photometric Tube Test Kit for Chromate
Catalog # MANA985024
Supplier:  MACHEREY-NAGEL
CAS Number:  
undefined
NANOCOLOR® Photometric Tube Test Kit for Chromate
Catalog # MANA985024
Supplier:  MACHEREY-NAGEL
CAS Number:  

Specifications

  • Chemical method:
    Carbazide
  • Pk:
    1 KIT
  • Description:
    NANOCOLOR® Chromate 5, test no. 0-24
  • Dimensions:
    160×110×119 mm (including packaging)
  • Model:
    MANA985024
  • Platform:
    NANOCOLOR® tube tests
  • Storage temperature:
    15…25 °C
  • Shelf life (months):
    24
  • Weight:
    315,6 g (including packaging)
  • For unit:
    UV/VIS II;VIS II;PF-12Plus;500 D;Advance;MT Easy VIS;MT Easy UV
  • Tests per kit:
    20
  • Range:
    0,05 - 2,00 mg/L; 0,1 - 4,0 mg/L (CrO₄²⁻)

Specifications

About this item

A pre-dosed NANOCOLOR® tube test for photometric determination of chromate in water.

  • Measurement directly in the test tube, saving time and reducing exposure to chemical reagents

  • Barcode cuvette identification
  • Precisely pre-dosed reagents in 16 mm (ext.Ø) tubes

The test is particularly suitable for NANOCOLOR® photometers but can be used with other photometers as well.

Sea water analysis: Yes

Hazardous material: No

Remark: Measuring range on NANOCOLOR VIS II

Delivery: Rugged box with 20 test tubes.