Orders LinkContactUsLinkComponent
Distributor for capillaries, b.safe
Distributor for capillaries, b.safe
Capillary connector, 3× capillary connector, 3× fittings for tubing Ext.Ø 3,2 mm, UNF 1/4"
Catalog # 590-0195
Supplier:   Bohlender
Distributor for capillaries, b.safe
Catalog # 590-0195
Supplier:  Bohlender
Distributor for capillaries, b.safe
Distributor for capillaries, b.safe
Capillary connector, 3× capillary connector, 3× fittings for tubing Ext.Ø 3,2 mm, UNF 1/4"
Price
51,60 €
Log in for availability
1 * 1 ST
Product Details & Documents
Distributor for connection of capillary tubing. With PFA Fittings for capillaries with Ext.Ø 3,2 mm (thread UNF ¹/₄").
  • Excellent chemical resistance
  • Available for a wide range of hose diameters
  • The conical shape gets the connection tight and leak-free
Adaptor to enlarge the number of capillary connectors on b.safe Caps.
Specifications
  • Thread:
    UNF 1/4"
  • With:
    3× capillary connector, 3× fittings for tubing Ext.Ø 3,2 mm
  • Description:
    Capillary connector