Order Entry
Austria
ContactUsLinkComponent
Suction filters with PTFE membrane, b.safe
Suction filters with PTFE membrane, b.safe
Catalog # 590-0221
Supplier:  Bohlender
undefined
Suction filters with PTFE membrane, b.safe
Catalog # 590-0221
Supplier:  Bohlender

Specifications

  • Ø:
    13 mm
  • Pore size:
    65 µm
  • Description:
    Suction filter with PTFE membrane
  • Pk:
    5
  • For tube O-Ø:
    3,2 mm (¹/₈") mm

Specifications

About this item

Suction Filter with PTFE membrane.

  • With a bore to be put on capillary tubing
  • Restrains pollution from solvents
  • For constant analysis results

Exchange the filter regularly every three months for reliable operation.