Orders LinkContactUsLinkComponent
Septum adaptor, b.safe
Septum adaptor, b.safe
Septum adapter for capillary connector UNF ¹/₄" to septum cap ND9
Catalog # 590-0182
Supplier:   Bohlender
Septum adaptor, b.safe
Catalog # 590-0182
Supplier:  Bohlender
Septum adaptor, b.safe
Septum adaptor, b.safe
Septum adapter for capillary connector UNF ¹/₄" to septum cap ND9
Price
33,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Threaded adaptor to connect a septum cap with a capillary connector of b.safe caps.
  • Easy sampling
  • Without contamination
  • Without evaporation
Easy sampling on the capillary connectors of b.safe caps and waste caps without contamination or evaporation of solvents.
Specifications
  • Description:
    Septum adapter for capillary connector UNF ¹/₄" to septum cap ND9