Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Fluoro-3-methylbenzo[b]thiophene
5-Fluoro-3-methylbenzo[b]thiophene 97%
5-Fluoro-3-methylbenzo[b]thiophene 97%
APOSPC9452-1G
:
Apollo Scientific
:
5-Fluoro-3-methylbenzo[b]thiophene 97%
APOSPC9452-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
68 °C/0.5mmHg
:
GHS07
:
Refrigerator
:
17514-63-5
:
C₉H₇FS
:
MFCD00179348