
2-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]sulphanyl}acetonitrile
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:306976-81-8
:
Formula:C₈H₄ClF₃N₂S
MDL Number:MFCD00215247
:252.65 g/mol
Melting Pt:64…66 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together