2-[3-Chloro-5-(trifluoromethyl)-2-pyridyl]-2-(4-methoxyphenyl)acetonitrile 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:338407-10-6
:
Formula:C₁₅H₁₀ClF₃N₂O
MDL Number:MFCD00172212
Melting Pt:100…102 °C
Storage Temperature:Ambient
Frequently Bought Together