
4,5-Dimethoxy-2-fluorobenzonitrile
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:119396-88-2
:
Formula:C₉H₈FNO₂
MDL Number:MFCD00143236
:181.1637 g/mol
Melting Pt:143 °C
Storage Temperature:Ambient
UN:3439
ADR:6.1,III
:GHS06
Frequently Bought Together