Order Entry
Austria
ContactUsLinkComponent
1,5-Dimethyl-3-(trifluoromethyl)-1H-pyrazole
Corrosive
1,5-Dimethyl-3-(trifluoromethyl)-1H-pyrazole
Catalog # APOSPC7948-1G
Supplier:  Apollo Scientific
CAS Number:  
Corrosive
1,5-Dimethyl-3-(trifluoromethyl)-1H-pyrazole
Catalog # APOSPC7948-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :164.13 g/mol
 :GHS05
Storage Temperature:Ambient
CAS Number:79080-31-2
Formula:C₆H₇F₃N₂
MDL Number:MFCD04972844