Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Tris[3,5-bis(trifluoromethyl)phenyl]phosphine
Tris[3,5-bis(trifluoromethyl)phenyl]phosphine 95%
Tris[3,5-bis(trifluoromethyl)phenyl]phosphine 95%
APOSPC7882-1G
:
Apollo Scientific
:
Tris[3,5-bis(trifluoromethyl)phenyl]phosphine 95%
APOSPC7882-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
670.28 g/mol
:
128…130 °C/0.5mmHg
:
98…102 °C
:
GHS07
:
Freezer
:
175136-62-6
:
C₂₄H₉F₁₈P
:
MFCD00042428