

2-Nitro-4-(trifluoromethyl)benzenesulphonyl chloride 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:837-95-6
:
Formula:C₇H₃ClF₃NO₄S
MDL Number:MFCD00013456
:289.62
Melting Pt:42…44 °C
Flash Pt:113 °C
Storage Temperature:Ambient
UN:3261
ADR:8,II
:GHS05
:GHS07
Frequently Bought Together