2,6-Dimethyl-4-[2-nitro-4-(trifluoromethyl)phenyl]morpholine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:405910-36-3
:
Formula:C₁₃H₁₅F₃N₂O₃
MDL Number:MFCD02018696
Melting Pt:61 °C
Storage Temperature:Ambient
Frequently Bought Together