
Methyl-2,5-bis(trifluoromethyl)-3,6-dioxaundecafluorononanoate (mixture of isomers) 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:26131-32-8
:
Formula:C₁₀H₃F₁₇O₄
MDL Number:MFCD00054723
Boiling Pt:155…156 °C
Density:1.66 g/cm³ (25 °C)
:510.10 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together