2'-Nitro-4'-(trifluoromethoxy)acetanilide 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:787-57-5
:
Formula:C₉H₇F₃N₂O₄
MDL Number:MFCD00052324
Melting Pt:94…96 °C
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together