
6-Chloro-2-fluoro-3-methoxybenzonitrile 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1017777-72-8
:
Formula:C₈H₅ClFNO
MDL Number:MFCD09832269
:185.59 g/mol
Melting Pt:88…92 °C
Storage Temperature:Ambient
UN:3439
ADR:6.1,III
:GHS07
Frequently Bought Together