2-Fluoropropane 98%
Catalog # APOSPC4179T-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Cylinder
- Pk:25 g
- Seq:3308
- Pk Info:1/4" NPT connection
Specifications
About this item
:
Boiling Pt:–10 °C
Melting Pt:–133.4 °C
Density:0.969 g/cm³ (25 °C)
ADR:2.1,
:62.09 g/mol
UN:3161
:GHS02
:GHS04
:GHS07
Storage Temperature:Ambient
CAS Number:420-26-8
Formula:C₃H₇F