5-Fluoro-2'-deoxyuridine (floxuridine) 99%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:50-91-9
:
Formula:C₉H₁₁FN₂O₅
MDL Number:MFCD00006530
:246.19 g/mol
Melting Pt:148…151 °C
Storage Temperature:Refrigerator
EINECS:200-072-5
UN:2811
ADR:6.1,III
:GHS06
:GHS08
Frequently Bought Together