3,5,5,6-Tetrafluoro-3,6-bis(trifluoromethyl)-1,4-dioxan-2-one 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:7309-84-4
:
Formula:C₆F₁₀O₃
MDL Number:MFCD04038350
:310.05 g/mol
Boiling Pt:70…70.5 °C
Storage Temperature:Ambient
Frequently Bought Together