Order Entry
Austria
ContactUsLinkComponent
2-(4-Chloro-2-(trifluoromethyl)phenyl)acetonitrile 98%
2-(4-Chloro-2-(trifluoromethyl)phenyl)acetonitrile 98%
Catalog # APOSPC302971-1G
Supplier:  Apollo Scientific
CAS Number:  
2-(4-Chloro-2-(trifluoromethyl)phenyl)acetonitrile 98%
Catalog # APOSPC302971-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :219.6 g/mol
Storage Temperature:Ambient
UN:3439
ADR:6.1,III
CAS Number:85386-80-7
Formula:C₉H₅ClF₃N
MDL Number:MFCD04115841