2,3-Difluoro-p-toluidine
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: 2,3-Difluoro-4-methylaniline
CAS Number:886503-79-3
:
Formula:C₇H₇F₂N
MDL Number:MFCD06660321
:143.14 g/mol
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together