3-(4-Fluorobenzyl)-5,5-dimethyl-1H-imidazole-2,4(3H,5H)-dione 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:860787-36-6
:
Formula:C₁₂H₁₃FN₂O₂
MDL Number:MFCD05975052
:236.25 g/mol
Storage Temperature:Ambient
Frequently Bought Together